Anti-ERP57 Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ADISPA585D ADISPA585F
ENZOADISPA585DEA 1800 DKK
ENZOADISPA585D ENZOADISPA585F
Anti-ERP57 Rabbit Polyclonal Antibody
Antistoffer
ERp57 (also known as Grp58), a member of the Thioredoxin superfamily, is an ER protein with disulfide isomerase activity. It has been shown to increase after oncogenic transformation and to be a component of the MHC Class I peptide-loading complex.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig and rabbit ERp57. Detects a band of ~57kDa by Western blot.

Type: Primary
Antigen: ERP57
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR