Anti-CRYAB Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ADISPA227D ADISPA227F
ENZOADISPA227DEA 2280 DKK
ENZOADISPA227D ENZOADISPA227F
Anti-CRYAB Rabbit Polyclonal Antibody
Antistoffer
α-Crystallins composed of αA (~19 kDa) and αB (~19.2 kDa) subunits, are major water-soluble proteins accounting for almost 50% of total protein in the mammalian transparent eye lens and they are also found in a variety of other tissues. Alpha-crystallins are also referred to as small heat shock proteins, since they are induced by increased temperature in a variety of organisms. The α-crystallins have sequence homology as well as structural and functional similarities with the small Hsp’s such as Hsp25/27. Most α-crystallins have four common structural and functional features: (i) molecular weight between 12 and 43kDa; (ii) the formation of large oligomeric complexes composed of αA-crystallin, αB-crystallin and Hsp25/27; (iii) the moderately conserved α-crystallin domain in the central region of the protein; and (iv) molecular chaperone activity. The α-crystallin domain comprises approximately 90 residues, is bounded by variable N-terminal and C-terminal extensions and is involved in oligomer assembly. Oligomers can reach 800 kDa or more and are dynamic, exhibiting subunit exchanges and organizational plasticity, possibly leading to functional diversity. Phosphorylation of serine residues occurs during development and in response to stress, and usually decreases oligomer size. Chaperone activity requires, and is modulated by, oligomerization and is limited to binding unfolded intermediates to prevent irreversible aggregation. Although productive release and refolding of denatured proteins requires close cooperation with other chaperones. Other proposed functions include a role in membrane stabilization and modulation of intermediate filament organization during physiological stress and neurodegenerative disease. Alpha B Crystallin is phosphorylated on serine 45 by ERK1/2 and serine 59 by MAPKAPK-2. When mono-phosphoylated αB-crystallin is isolated it contains only αB-crystallin phosphorylated at serine 19.

  • For Immunoprecipitation, Western Blot

Recognizes mouse, rat, bovine, pig and sheep alphaB-crystallin.

Type: Primary
Antigen: CRYAB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR