Anti-CRYAB Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ADISPA225C ADISPA225E
ENZOADISPA225CEA 1590 DKK
ENZOADISPA225C ENZOADISPA225E
Anti-CRYAB Rabbit Polyclonal Antibody
Antistoffer
Alpha-crystallins, which are part of the small Heat shock family members, are major water-soluble proteins present in the lens of the mammalian eye. Phosphorylation of serine residues which occurs during development and in response to stress, is intimately linked with its function. Chaperone activity requires, and is modulated by, oligomerization and is limited to binding unfolded intermediates to prevent irreversible aggregation.

  • For Immunoprecipitation, Western Blot

Recognizes human, mouse and bovine alphaB-crystallin.

Type: Primary
Antigen: CRYAB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR