Anti-SOD2 Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ADISOD111D ADISOD111F
ENZOADISOD111DEA 2530 DKK
ENZOADISOD111D ENZOADISOD111F
Anti-SOD2 Rabbit Polyclonal Antibody
Antistoffer
SOD (Superoxide dismutase) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells, they are: EC SOD (extracellular SOD), Cu/Zn SOD (copper and zinc-containing SOD) and Mn SOD (manganese-containing SOD).

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, bovine, chicken, clam, dog, Drosophila, guinea pig, hamster, monkey, pig, rabbit, sheep, and Xenopus Mn SOD. Detects a band of ~25kDa by Western blot.

Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR