Anti-TCP1A Rat Monoclonal Antibody
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ADICTA191D ADICTA191F
ENZOADICTA191DEA 2060 DKK
ENZOADICTA191D ENZOADICTA191F
Anti-TCP1A Rat Monoclonal Antibody
Antistoffer
TCP-1 alpha is a 60 kDa subunit of a cytosolic hetero-oligomer chaperone that is known to be involved in the folding of Actin and Tubulin. This protein is a member of the Chaperonin family, which includes E. coli GroEL, the mitochondrial Heat-shock protein Hsp60, the plastid Rubisco-subunit-binding protein, and the archaebacterial protein TF55. These Chaperonins assist the folding of proteins upon ATP hydrolysis.
  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse, rat, bovine, dog, C. elegans,More Product Information