DCpak® SFC Achiral (SFC- A) Columns
Leverandør: CHIRAL TECHNOLOGIES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AA424 AA425 AA435 AA445 AA494 AA495 AAS82 AAS83 AAS84 AAS92 AAS93 AAS94
DAICAA424EA 7470 DKK
DAICAA424 DAICAA425 DAICAA435 DAICAA445 DAICAA494 DAICAA495 DAICAAS82 DAICAAS83 DAICAAS84 DAICAAS92 DAICAAS93 DAICAAS94
DCpak® SFC Achiral (SFC- A) Columns
Kromatografikolonner
When a material is in a state that the temperature and the pressure exceed the critical point, it becomes a supercritical fluid. In this phase, the density of gas and liquid becomes the same.