Bags, radiation bin box, B-Safe
Leverandør: Alpha Packaging
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
526081 526083 526084 526085
129-0407EA 2040 DKK
129-0407 129-0409 129-0410 129-0411
Bags, radiation bin box, B-Safe
Poser Affaldsposer
The bags are printed with yellow radioactive warning symbol and a maximum fill line. PE,125 µm (500 gauge), clear.
  • Heat sealed for leak protection
  • Each bag is box-shaped and has a double-string neck-pull