Erlenmeyer flasks with joint, quartz
Leverandør: TECHNICAL QUARTZ SOLUTIONS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
75550 75551 75552 75553
TSLA75550EA 4520 DKK
TSLA75550 TSLA75551 TSLA75552 TSLA75553
Erlenmeyer flasks with joint, quartz
Kolber Erlenmeyer-kolber
Made of quartz glass, clear, for application temperature up to 1050 °C.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes