Disposal Bags, PP
Leverandør: SEMADENI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7757 7755 7756
SEMA7757EA 1650 DKK
SEMA7757 SEMA7755 SEMA7756
Disposal Bags, PP
Poser Affaldsposer
These disposal bags are made of particularly strong foil, 50 μm, with wide bottom seam. For disposal of contaminated articles and laboratory waste.
Emballering: Packed in practical dispenser boxes.