CF™ azide reactive dyes
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
92180 92181 92182
92180.EA 3620 DKK
92180. 92181. 92182.
CF™ azide reactive dyes
Proteinkoblings- og mærkningssystemer
CF™ dye azide reacts with alkyne to form 1,2,3-triazole by copper(I) catalysed 1,3-dipolar Huisgen cycloaddition. It could also undergo Staudinger ligation with a phosphine containing compound.