Nitrile gloves, Showa 707D
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
707D-07 707D-08 707D-11
112-5887EA 261 DKK
112-5887 112-5888 112-5891
Nitrile gloves, Showa 707D
Handsker
This hybrid nitrile glove offers protection against chemicals while also being approved for handling foodstuffs.
  • Thin, light, unsupported glove with a "second skin" feel
  • Protects the hand from oils, hydrocarbons, grease and abrasion
  • Embossed finish with tractor-tread pattern: Good grip
  • Impermeable for working in damp or greasy environments
  • Cuff prevents dirt from entering the glove
  • Easy to put on and remove, lint-free, dust-free
  • Semi-disposable; can be used once or re-used
Typical applications include: Automotive, chemical, food, manufacturing, pharmaceutical.
Certificeringer: EN ISO 374-1 Type A (JKLOPT), EN ISO 374-5; Cat. III
Food contact approved