Gemfibrozil 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: CI-719

461790050 461790250
ACRO461790050EA 532 DKK
ACRO461790050 ACRO461790250
Gemfibrozil 98%
Gemfibrozil
Formel: C₁₅H₂₂O₃
Smeltepunkt: 60…61 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00079335
CAS-nummer: 25812-30-0

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 58 to 63 °C
Water ≤0.25 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR