High performance centrifuge bottles, Sorvall™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
010-1456 010-1459
SORV010-1456EA 0 DKK
SORV010-1456 SORV010-1459
High performance centrifuge bottles, Sorvall™
Flasker Centrifugeflasker
These bottles are ideal for bacterial, yeast tissue isolations and viral harvesting.
  • High-strength aluminum cap enhances bottle support
  • Improves chemical resistance to phenol solutions
Smooth inner bottle walls designed to reduce shearing in mammalian culture separations.
Polypropylene copolymer (PPCO) bottles are designed for excellent chemical resistance and are autoclavable.
Polycarbonate (PC) bottles designed for excellent clarity and mechanical strength.