Jars, wide mouth, with pressure self seal cap
Leverandør: LAMAPLAST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2434-1 2430-1 2426-1 2420-1 2418-1 2415-1 2433-1
215-3723EA 2440 DKK
215-3723 215-3719 215-3717 215-3715 215-3713 215-3699 LAMA2433-1
Jars, wide mouth, with pressure self seal cap
Dåser Dåser, allround
These wide mouth jars are made of PP. Suitable to storage: creams, semi-solid substances, crystals, granulates and powders.
  • Self-sealing screw cap
  • Guaranteed the tightness of the content once you break the seal
  • Can be closed again after the first time by pushing the cap on the mouth
  • With good chemical and temperature resistantance
Colour: White
Certificeringer: Meets 1935/2004/CE and UE 10/2011.
Bestillingsoplysninger: Caps must be ordered separately.