Dishes, evaporating, french shape
Leverandør: MORGAN ADVANCED MATERIALS HALD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L130/5 L130/6 L130/7
HALDL130/5EA 432 DKK
HALDL130/5 HALDL130/6 HALDL130/7
Dishes, evaporating, french shape
Bakker/Tallerkener/Potter Fordampningsbassiner/skåle
Porcelain, glazed.
  • Very good chemical resistance
  • Heat resistant up to +1000 °C
  • Low thermal expansion
French form, with spout and round bottom, glazed.