Tris-HCl, 1M solution, pH 8,0, Molecular Biology Grade, Ultrapure
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J22638.AE J22638.AP
J22638.AEEA 487 DKK
J22638.AE J22638.AP
Tris-HCl, 1M solution, pH 8,0, Molecular Biology Grade, Ultrapure
Buffere Kloning buffere
1M solution of Tris, Ultrapure in high purity distilled H₂O, adjusted to pH 8,0.