Cut resistant gloves, uvex C500 wet plus
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6049611 6049610 6049609 6049608 6049607
UVEX6049611EA 198 DKK
UVEX6049611 UVEX6049610 UVEX6049609 UVEX6049608 UVEX6049607
Cut resistant gloves, uvex C500 wet plus
Handsker
These bamboo-viscose/glass/HPPE/polyamide gloves feature a HPE coating on palm and fingers. The Bamboo TwinFlex® technology ensures maximum cut resistance, exceptional abrasion resistance and outstanding wearing comfort. Suitable for damp, oily or greasy applications.
  • Resistant to contact heat up to 100 °C
  • Oil and grease resistant
  • Outstanding wearing comfort and tactile feel with climazone technology
  • Very good grip in damp and wet environments
  • Highest cut resistance level 5
  • Silicone-free according to imprint test
Typical applications include: automotive industry, construction, brewery/beverage industry, glass industry, maintenance/servicing, metal industry, assembly works, paper industry, shipping/logistics, transportation works.
Certificeringer: EN 388:2016 (4X42C)
OEKO-TEX® Standard 100