UHPLC Purospher® STAR RP-18 endcapped kolonner
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.50650.0001 1.50651.0001 1.50645.0001 1.50646.0001 1.50654.0001 1.50648.0001 1.50653.0001 1.50649.0001 1.50648.1003
1.50650.0001EA 4210 DKK
1.50650.0001 1.50651.0001 1.50645.0001 1.50646.0001 1.50654.0001 1.50648.0001 1.50653.0001 1.50649.0001 1.50648.1003
UHPLC Purospher® STAR RP-18 endcapped kolonner
Kromatografikolonner
Available with optimised 2 μm particulate silica, these UHPLC columns, based on Purospher® STAR RP-18 endcapped, are ideal for ultra-fast applications, where resolution, sensitivity and sample throughput are crucial. They are the first choice for high throughput screening and QC analyses, process monitoring, method development and LC-MS applications. Due to its balanced selectivity, Purospher® STAR RP-18 endcapped UHPLC columns cover almost all demanding separations with tailing-free chromatograms. Purospher® STAR RP-18 endcapped 3 μm columns are recommended for difficult samples where clogging and back pressure present an issue.
  • In UHPLC environment, speed is increased by a factor of 10
  • Solvent consumption is drastically cut
  • Excellent peak shape for all types of acidic, basic and metal-chelating analytes
  • Extraordinary pH stability from pH 1,5 to 10,5 for an extremely wide application range
  • Enhanced sensitivity due to improved signal-to-noise ratio
Hibar® HR columns are complete with end fittings and pressure stable (600 bar) for UHPLC use.
Hibar® HR columns are complete with end fittings and pressure stable (600 bar) for UHPLC use.