Colistin sulphate
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
455390010 455392500
ACRO455390010EA 1630 DKK
ACRO455390010 ACRO455392500
Colistin sulphate
Proteiner
  • May contain up to 15% water
  • HPLC: ≥77 % Sum of polymyxins