Accessories for comparators
Leverandør: Lovibond
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
221270 230160 231200 246410 246420 246430 246440 246450 243030 243040 221090 230080 230120 230130 230250 230460 234050 235010 235020 235330 237200 237710 231070 281030 281040 281060
LOVI221270EA 2280 DKK
LOVI221270 LOVI230160 LOVI231200 TINT246410 TINT246420 TINT246430 TINT246440 TINT246450 TINT243030 TINT243040 LOVI221090 LOVI230080 LOVI230120 LOVI230130 LOVI230250 LOVI230460 LOVI234050 LOVI235010 LOVI235020 LOVI235330 LOVI237200 LOVI237710 LOVI231070 LOVI281030 LOVI281040 LOVI281060
Accessories for comparators
Kolorimetre Komparator-sæt
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+