Pipe blind lockouts
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
149228 149229 149230
BRDY149228EA 811 DKK
BRDY149228 BRDY149229 BRDY149230
Pipe blind lockouts
Sikkerhedslåse
These devices can be applied to the widest range of pipes to easily and temporarily avoid the release of gas, steam and hazardous liquids while pipe maintenance is still ongoing.
  • Adjustable locking arm accommodates both standard and recessed bolt positions
  • Secure 1 or 2 bolts for extra security
  • Heavy duty construction withstands harsh environments
  • UL approved for use in explosive atmospheres
Min. shackle clearance: 19,05 mm
Max. shackle diameter: 7,00 mm