Monensin monosodium salt 90%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

461450010 461450050 461450250
ACRO461450010EA 743 DKK
ACRO461450010 ACRO461450050 ACRO461450250
Monensin monosodium salt 90%
Monensin monosodium salt
Formel: C₃₆H₆₁NaO₁₁
Smeltepunkt: 267…269 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00077826
CAS-nummer: 22373-78-0
UN nummer: 2811
ADR: 6.1,II

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Powder
Infrared spectrum Conforms
Solubility (5 % in Methanol) Clear colorless to light yellow
HPLC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR