Abacavir sulphate 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: ((1S,4R)-4-(2-Amino-6-(cyclopropylamino)-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol hemisulphate

458860010 458860050
ACRO458860010EA 500 DKK
ACRO458860010 ACRO458860050
Abacavir sulphate 99%
Abacavir sulphate
Formel: C₂₈H₃₈N₁₂O₆S
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04112763
CAS-nummer: 188062-50-2

Bestil nu

Specifikationer Testresultater

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR