Indomethacin

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

458030050 458030250 458031000
ACRO458030050EA 336 DKK
ACRO458030050 ACRO458030250 ACRO458031000
Indomethacin
Indomethacin
Formel: C₁₉H₁₆ClNO₄
Molekylvægt: 357.80 g/mol
Smeltepunkt: 155 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00057095
CAS-nummer: 53-86-1
UN nummer: 2811
ADR: 6.1 II

Bestil nu

Specifikationer Testresultater

Appearance White to yellow to light beige Crystalline powder (May have a pink hue)
Infrared spectrum Conforms
Melting point 156 to 165 °C
Loss on drying ≤0.5 % (100 °C, 2 h, 5 mm Hg)
HPLC ≥97.5 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR