6-Heptynoic acid 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
457390010
457390050
457390250
ACRO457390010EA
358
DKK
ACRO457390010
ACRO457390050
ACRO457390250
6-Heptynoic acid 95%
6-Heptynoic acid
Formel:
C₇H₁₀O₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00239430 CAS-nummer: 30964-00-2 UN nummer: 3265 |
Specifikationer Testresultater
Appearance | Colorless to yellow Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...