Eddikesyre 50% in water
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
456870050 456870250
ACRO456870050EA 491 DKK
ACRO456870050 ACRO456870250
Eddikesyre 50% in water
Eddikesyre
Formel: H₃CCOOH
Molekylvægt: 60.05 g/mol
Kogepunkt: 118
Smeltepunkt: 17
Flammepunkt: >60 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00036152
CAS-nummer: 64-19-7
UN nummer: 2790
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Clear colorless Solution
Titration with NaOH 49.0 to 51.0 %