Chondroitin sulphate sodium salt

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

456280010 456280050 456280250
ACRO456280010EA 529 DKK
ACRO456280010 ACRO456280050 ACRO456280250
Chondroitin sulphate sodium salt
Chondroitin sulphate sodium salt
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00130792
CAS-nummer: 9082-07-9

Bestil nu

Specifikationer Testresultater

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤12 % (105°C, 4 h)
Residue after ign. (sulfated) 15 % to 30 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR