2-Chloro-4-ethynylpyridine 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

449980010 449980050
ACRO449980010EA 1630 DKK
ACRO449980010 ACRO449980050
2-Chloro-4-ethynylpyridine 97%
2-Chloro-4-ethynylpyridine
Formel: C₇H₄ClN
Kogepunkt: 373 °C (760 mmHg)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13189742
CAS-nummer: 945717-09-9

Bestil nu

Specifikationer Testresultater

Appearance White to light brown to green Crystalline powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR