Calciumcarbonat ACS chelometric standard

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Marble

403801000 403805000
ACRO403801000EA 339 DKK
ACRO403801000 ACRO403805000
Calciumcarbonat ACS chelometric standard
Calcium carbonate
Formel: CaCO₃
Molekylvægt: 100.09 g/mol
Smeltepunkt: 825 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010906
CAS-nummer: 471-34-1
Merck Index: 13,01658

Bestil nu

Specifikationer Testresultater

Appearance White Crystalline powder
Titration Complexometric 99.95 to 100.05 % (on dried substance)
Chloride (Cl) ≤0.001 %
Fluoride (F) ≤0.0015 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.003 %
Barium (Ba) ≤0.01 %
Iron (Fe) ≤0.002 %
Magnesium (Mg) ≤0.01 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.01 %
Strontium (Sr) ≤0.1 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.01 % (in dilute HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR