(±)-4-Phenyl-2-butanol 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

183400250 183400050
ACRO183400250EA 877 DKK
ACRO183400250 ACRO183400050
(±)-4-Phenyl-2-butanol 97%
(±)-4-Phenyl-2-butanol
Formel: (C₆H₅)CH₂CH₂CH(OH)CH₃
Kogepunkt: 132 °C (14 mmHg)
Massefylde: 0,98 g/cm³
Flammepunkt: >110 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00044349
CAS-nummer: 2344-70-9

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5150 to 1.5180 (20°C, 589 nm)
GC ≥96.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR