Chemical resistant overalls, 9877
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9871011 9871013 9871010 9871014 9871012
UVEX9871011EA 4030 DKK
UVEX9871011 UVEX9871013 UVEX9871010 UVEX9871014 111-2614
Chemical resistant overalls, 9877
Engangsbeklædning
These disposable overalls are made of a very light, microporous PE laminate, combined with an SMS back section for high breathability. The overalls guarantee a high level of protection against liquid splashes and solid particles.
  • Particle-tight and spray-tight to a limited extent
  • Free of silicone and paint wetting impairment substances
  • Bound seams in contrasting colours for visible safety
  • Middle finger loops prevent sleeves sliding up the arm
  • Self-adhesive zipper flap for optimal protection
  • Optimal fit with elasticated waistband and elasticated bands on hood, arms and legs
  • Anti-static properties
Typical applications include: Handling chemicals in dust and powder form, varnishing and protection against paint splashes, fibreglass production and processing, industrial cleaning and maintenance, automotive industry, grinding and polishing, working with asbestos, wood and metal processing, construction industry, cleanroom applications, pharmaceutical industry.
Suitable for cleanroom environments Class 8 in accordance with ISO 14644-1.
Certificeringer: Type 5/6, EN 1149-5; Cat. III
UV protection factor UPF 50+