Dental micro spatulas, double-ended, Aesculap®
Leverandør: Aesculap
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DE440R DE442R DE443R DE444R DE445R
AESCDE440REA 0 DKK
AESCDE440R AESCDE442R AESCDE443R AESCDE444R AESCDE445R
Dental micro spatulas, double-ended, Aesculap®
Spatler
Micro spatulas for application of the retrograde filling material (Super EBA) into the dental cavity.