Table top combined refrigerators/freezers
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TX1021 KTS102 KTS126 KTS148 KTS165
471-1014EA 5480 DKK
471-1014 471-0993 471-0994 471-0995 LIEBKTS165
Table top combined refrigerators/freezers
Køleskabe
Table top refrigerators for general use.
  • Static cooling
  • Automatic defrost
  • 'EasyClean' interior provides full modularity and ultra-smooth walls for easy cleaning
  • Adjustable feet
  • Reversible door
  • Glass shelves
Temperature range: −3 to +8 °C