Reference standards, waste water - trace metals
Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SPS-WW1 SPS-WW2
PROOSPS-WW1EA 3150 DKK
PROOSPS-WW1 PROOSPS-WW2
Reference standards, waste water - trace metals
Standarder Miljø analyse standarder
This Reference material is intended for daily use in conjunction with measurement of elements in wastewaters. The water solution contains 13 elements with concentrations traceable to ultra pure metals or stoichiometrically well-defined substances.