Reference Materials, Matrix Modifiers for GFAA, CRM, VHG, Promochem
Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VHG-MMGN1P-100 VHG-MPM1-250 VHG-MPM2-250 VHG-MPM3-250
PROOVHG-MMGN1P-100EA 768 DKK
PROOVHG-MMGN1P-100 PROOVHG-MPM1-250 PROOVHG-MPM2-250 PROOVHG-MPM3-250
Reference Materials, Matrix Modifiers for GFAA, CRM, VHG, Promochem
Standarder Kationiske standarder Multi-Element Standards
This solution is intended for use as a matrix modifier for graphite furnace atomic absorption spectroscopy (GFAA) in the determination of trace elements.
Certificeringer: Traceable to NIST, SRM.