Multi-element standards, EPA calibration standards, CRM, VHG
Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VHG-44CS1Y-100 VHG-44CS1Y-500 VHG-44CS2Z-100 VHG-44CS2Z-500
PROOVHG-44CS1Y-100EA 3440 DKK
PROOVHG-44CS1Y-100 PROOVHG-44CS1Y-500 PROOVHG-44CS2Z-100 PROOVHG-44CS2Z-500
Multi-element standards, EPA calibration standards, CRM, VHG
Standarder Kationiske standarder Multi-Element Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma mass spectrometry (ICP-MS), or alternative techniques, such as inductively coupled plasma optical emission spectroscopy (ICP-OES), and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Certificeringer: Traceable to NIST, SRM.