Melting point standard, benzophenone, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
50296-1G 50296-5G
50296-1GEA 223 DKK
50296-1G 50296-5G
Melting point standard, benzophenone, Supelco®
Standarder Smeltepunkts Standarder
Melting point standards are used to identify compounds and estimate purity. These standards can be used with any melting point apparatus.
  • Ready to use
  • Traceable to primary reference material
  • Uncertainty of measurement up to ±0,3 °C
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1116c712c225cae7e9e35b3c1815aca674f6e2b2&vid=1116c712c225cae7e9e35b3c1815aca674f6e2b2&prid=web&clid=SAPDAM