Silver standard solution, for ICP, in HNO₃, CRM, VHG
Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VHG-PAGN-100 VHG-PAGN-500 VHG-TAGN-100 VHG-TAGN-500 VHG-LAGN-100
PROOVHG-PAGN-100EA 620 DKK
PROOVHG-PAGN-100 PROOVHG-PAGN-500 PROOVHG-TAGN-100 PROOVHG-TAGN-500 PROOVHG-LAGN-100
Silver standard solution, for ICP, in HNO₃, CRM, VHG
Standarder Kationiske standarder Silver Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificeringer: Traceable to NIST, SRM.