Biobanking tubes, Cryo.s™
Leverandør: Greiner Bio-One
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
978561_960 978570_960 978580_960 976561_960 976570_10 976567_960 977561_960 977570_960 977580_960 976580_960
479-2162EA 11370 DKK
479-2162 479-2163 479-2164 GREI976561_960 GREI976570_10 GREI976567_960 GREI977561_960 GREI977570_960 GREI977580_960 GREI976580_960
Biobanking tubes, Cryo.s™
Hætteglas
Cryo.s™ biobanking tubes are used for storage of tissue, cells, fungi, bacteria, spores, cellular extracts or body fluids at ultra-low temperature for research and development purposes in a very compact format.
  • Pre-racked
  • Non-capped tubes are non-sterile
  • Conserves upto 30% freezer space
Advarsel: Must not be stored in the liquid phase of liquid nitrogen, but only in the gas phase. Not intended for any application in the context of reproductive medicine.