Metallo-organic standards, V21+K wear metals standard, VHG
Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VHGV21K50-200G VHGV21K100100G VHGV21K50-100G VHGV21K100200G VHGV21K100400G VHGV21K500100G VHGV21K500200G VHGV21K10-100G
PROOVHGV21K50-200GEA 2840 DKK
PROOVHGV21K50-200G PROOVHGV21K100100G PROOVHGV21K50-100G PROOVHGV21K100200G PROOVHGV21K100400G PROOVHGV21K500100G PROOVHGV21K500200G PROOVHGV21K10-100G
Metallo-organic standards, V21+K wear metals standard, VHG
Standarder Kationiske standarder Multi-Element Standards
This standard solution is intended for use as a certified reference material and calibration standard for the analysis of these elements in petroleum products or other organic matrices using inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), rotating disc electrode atomic emission spectroscopy (RDE-AES), flame or furnace atomic absorption spectroscopy (AA or GFAA) and other techniques for elemental analysis.
Multi element standards for Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V and Zn in hydrocarbon oil.
Certificeringer: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.