Ruthenium(III) chloride oxide ≥85%, pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ruthenium oxychloride, Tetraamminechlorohydroxoruthenium(III) chloride, Ruthenium Red

279710010 279710050
ACRO279710010EA 1060 DKK
ACRO279710010 ACRO279710050
Ruthenium(III) chloride oxide ≥85%, pure
Ruthenium(III) chloride oxide
Formel: H₄₂Cl₆N₁₄O₂Ru₃
Molekylvægt: 786.354 g/mol
Smeltepunkt: >500 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00011479
CAS-nummer: 11103-72-3

Bestil nu

Specifikationer Testresultater

Appearance Dark red or purple Powder
Elemental Analysis 34 to 39 % (Ru)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR