Biodiesel standards, sulphur in B100 biodiesel, VHG
Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VHGSB10010-100 VHGSB100-5-100 VHGSB100100100
PROOVHGSB10010-100EA 2000 DKK
PROOVHGSB10010-100 PROOVHGSB100-5-100 PROOVHGSB100100100
Biodiesel standards, sulphur in B100 biodiesel, VHG
Standarder Kationiske standarder Sulphur Standards
This standard solution is intended for use as a certified reference material and calibration standard for the analysis of sulphur in B100 biodiesel using x-ray fluorescence spectrometry (XRF) and other techniques for sulphur analysis.
It is suitable for use with methods such as ASTM D2622, D4294, D5453, D6751 and others.
Certificeringer: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.