Vand, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Hydrogen oxide
Management of Change (MOC) category = NI
4218.2500EA 368 DKK
4218.2500 4218.1000
Vand, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Vand
Formel: H₂O
Molekylvægt: 18.02 g/mol
Kogepunkt: 100
Smeltepunkt: 0
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011332
CAS-nummer: 7732-18-5
Merck Index: 13,10098
Bestil nu
Specifikationer Testresultater
Particulate Matter max. 0.0001 %
Residue after Evaporation max. 2 ppm
Ultraviolet absorbance of largest eluted peak
at 220 nm max. 0.005 au
at 254 nm max. 0.001 au
Physical Data (not specifications):
Learn more