Sealing film for manual application, Parafilm® M
Leverandør: HEATHROW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HS234526C HS120265 HS234526A HS234526B
291-1212EA 960 DKK
291-1212 291-1211 291-1214 291-0057
Sealing film for manual application, Parafilm® M
Forsegling Plastfolie
Parafilm® M is a semi-transparent, flexible, thermoplastic film with two-way stretch. This odourless and colourless film provides a tight, moisture-proof seal.
  • Non-toxic and free from plasticisers
  • Primarily composed of polyolefins and paraffin waxes offering good resistance against acids, alkalis and some organic solvents*
  • Will stretch 200% and adhere to irregular shapes and surfaces to provide a secure seal
  • Disposable
Parafilm® is a registered trademark of Bemis Company, Inc.
Advarsel: *Use not recommended with acetone, diethyl ether or tetrachloride