Measuring scoops
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1900A-01 1900A-05 1900A-10 1900A-25 1900A-50
SAMP1900A-01EA 823 DKK
SAMP1900A-01 SAMP1900A-05 SAMP1900A-10 SAMP1900A-25 SAMP1900A-50
Measuring scoops
Øser
These measuring scoops made from high quality 316 stainless steel are ideal for scooping out fixed volumes of powder.
  • Crevice-free
  • Polished