Chloride ion chromatography standards, (from KCl), CRM, VHG
Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VHG-I1PCL-100 VHG-ICL1K-100 VHG-ICL1K-500 VHG-I1PCL-500
PROOVHG-I1PCL-100EA 1120 DKK
PROOVHG-I1PCL-100 PROOVHG-ICL1K-100 PROOVHG-ICL1K-500 PROOVHG-I1PCL-500
Chloride ion chromatography standards, (from KCl), CRM, VHG
Standarder Anioniske standarder Chloride Standards
This solution is intended for use as a certified reference material or calibration standard, for ion chromatography (IC) or techniques using other modes of aqueous ion detection.
Certificeringer: Traceable to NIST and SRM.