VWR®, Incineration Bags
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
129-1567EA 1650 DKK
129-1567 129-1569
VWR®, Incineration Bags
Poser Affaldsposer
Constructed of durable PP with feature the universal biohazard symbol in black.
Certificeringer: Meets the 165 g dart drop standard performed per ASTM D1709-98 and the 480 g tear standard performed per ASTM D1299-94A.
Advarsel: Autoclaving not recommended. Bags will melt at 98,9 °C.