pH indicator strips, Whatman™
Leverandør: Whatman products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10362000 10362010
700-0442EA 211 DKK
700-0442 WHAT10362010
pH indicator strips, Whatman™
Prøvepapir pH-indikatorstrips
Universal indicator strips which have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular colour. A check against the colour comparision table supplied allows the pH to be determined.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use