Air line regulator lockouts
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
064539 064540
BRDY064539EA 262 DKK
BRDY064539 BRDY064540
Air line regulator lockouts
Sikkerhedslåse
These air line regulator lockouts are designed to help users work safely when performing maintenance and repairing pneumatic systems.
  • Eliminates the problem of disabling other equipment sharing same air supply
  • Device regulator to be locked in the ON, OFF or THROTTLED position
  • Installation is as easy as removing the snap cap, installing the locking device on the regulator rod and snapping the replacement cap on the regulator
  • Glass-filled nylon body and aluminium stem
  • One padlock can be applied
Installation is as easy as removing the snap cap, installing the locking device on the regulator rod and snapping the replacement cap on the regulator.
Leveringsinformation: Supplied with lockout, cap and allen key.