ESD safety shoes for ladies, lace-up, New High Lady Black
Leverandør: MILLE S.A.S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NFHN3/38 NFHN3/37 NFHN3/39 NFHN3/40 NFHN3/41 NFHN3/42 NFHN3/36
MILENFHN3/38EA 877 DKK
MILENFHN3/38 MILENFHN3/37 MILENFHN3/39 MILENFHN3/40 MILENFHN3/41 MILENFHN3/42 MILENFHN3/36
ESD safety shoes for ladies, lace-up, New High Lady Black
Sko og støvler
These anti-static, metal-free safety shoes are made from full grain leather upper plus an extra light, PU, dual-density outer sole.
  • Abrasion resistant, antibacterial, poromax® lining giving high water-absorbing capacity for additional breathability
  • Waterproof tongue and heel grip
  • Textile, puncture resistant midsole (1430 N)
  • Toe protection cap (240 J) made from fibre glass insulates from warmth and cold
Suitable for industry, transport, logistics and services.
Colour: Black
Certificeringer: EN ISO 20345; IEC EN 61340-5-1 (ESD); S3, SRC.